| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015277.hg.1 |
ENST00000265381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015277.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015277.hg.1 |
TC09001193.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900170824.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900170824.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900170824.hg.1 |
ENST00000265381.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900170824.hg.1 |
NM_001163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900170824.hg.1 |
TC0900010350.hg.1 |
Clariom D TC |