| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056860.hg.1 |
ENST00000367267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056860.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056860.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056860.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056860.hg.1 |
TC01003705.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228418.hg.1 |
TC0100016960.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
ENST00000367268.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_001136504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016960.hg.1 |
NM_177402.4 |
Refseq |