| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02017408.hg.1 |
ENST00000428675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02017408.hg.1 |
ENST00000437704.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02017408.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02017408.hg.1 |
TC02001152.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224636.hg.1 |
TC0200016587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000428675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000435320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000437704.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
ENST00000487695.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016587.hg.1 |
NM_006226.3 |
Refseq |