| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029588.hg.1 |
ENST00000258243.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029588.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01029588.hg.1 |
TC01001905.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100187893.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100187893.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100187893.hg.1 |
ENST00000258243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100187893.hg.1 |
NM_014777.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100187893.hg.1 |
TC0100011913.hg.1 |
Clariom D TC |