Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00987955_g1
          See other CAST GE Assays ›
          Gene Symbol:
          CAST
          Gene Name:
          calpastatin
          Gene Aliases:
          BS-17, PLACK
          Chromosome Location:
          Chr.5: 96662037 - 96774683 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00987955_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CAST
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190442.1 NP_001177371.1 7 - 8 487 97
          NM_001284213.2 NP_001271142.1 4 - 5 284 97
          NM_173060.3 NP_775083.1 4 - 5 284 97
          XM_006714696.3 XP_006714759.1 8 - 9 590 97
          XM_006714697.3 XP_006714760.1 7 - 8 607 97
          XM_006714698.3 XP_006714761.1 7 - 8 607 97
          XM_006714699.3 XP_006714762.1 7 - 8 531 97
          XM_006714701.3 XP_006714764.1 6 - 7 566 97
          XM_006714703.3 XP_006714766.1 6 - 7 566 97
          XM_006714704.3 XP_006714767.1 6 - 7 475 97
          XM_006714707.3 XP_006714770.1 5 - 6 353 97
          XM_006714708.3 XP_006714771.1 5 - 6 352 97
          XM_006714710.3 XP_006714773.1 4 - 5 270 97
          XM_011543654.2 XP_011541956.1 7 - 8 607 97
          XM_011543655.2 XP_011541957.1 7 - 8 607 97
          XM_011543656.2 XP_011541958.1 6 - 7 551 97
          XM_011543657.2 XP_011541959.1 6 - 7 433 97
          XM_011543658.2 XP_011541960.1 4 - 5 286 97
          XM_017009911.1 XP_016865400.1 7 - 8 607 97
          XM_017009912.1 XP_016865401.1 8 - 9 618 97
          XM_017009915.1 XP_016865404.1 7 - 8 552 97
          XM_017009916.1 XP_016865405.1 6 - 7 566 97
          XM_017009917.1 XP_016865406.1 7 - 8 523 97
          XM_017009919.1 XP_016865408.1 7 - 8 607 97
          XM_017009920.1 XP_016865409.1 6 - 7 433 97
          XM_017009921.1 XP_016865410.1 6 - 7 450 97
          XM_017009927.1 XP_016865416.1 5 - 6 352 97
          XM_017009928.1 XP_016865417.1 5 - 6 390 97
          XM_017009929.1 XP_016865418.1 4 - 5 270 97
          XM_017009931.1 XP_016865420.1 4 - 5 270 97
          GenBank mRNA AB208816.1 - 7 - 8 451 97
          AK296919.1 - 6 - 7 406 97
          AK297879.1 - 5 - 6 265 97
          AK298541.1 - 5 - 6 353 97
          AK299384.1 - 5 - 6 257 97
          AK299402.1 - 5 - 6 333 97
          AK300624.1 - 4 - 5 267 97
          AK303900.1 - 4 - 5 271 97
          AK314290.1 - 4 - 5 268 97
          AK315146.1 - 4 - 5 97
          AL832174.1 - 4 - 5 269 97
          AL832349.1 - 4 - 5 272 97
          BC016066.1 - 6 - 7 587 97
          BQ420953.1 - 5 - 6 333 97
          CD702474.1 - 5 - 6 384 97
          D16217.1 - 5 - 6 346 97
          DC424542.1 - 4 - 5 284 97
          M86258.1 - 4 - 5 184 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500013204.hg
          This assay targets the same location as HTA transcript cluster(s) TC05000469.hg

          Additional Information:

          This assay is in the 5'UTR region of the AK315146.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CAST

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          egg activation
          brain development
          myoblast fusion
          aging
          negative regulation of endopeptidase activity
          organ regeneration
          myoblast differentiation
          negative regulation of cell cycle arrest
          cell-cell adhesion
          negative regulation of type B pancreatic cell apoptotic process
          protease binding
          endopeptidase inhibitor activity
          protein binding
          calcium-dependent cysteine-type endopeptidase inhibitor activity
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000309190.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000325674.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000338252.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000341926.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000348386.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000359176.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000395812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000395813.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000504465.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000508608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000508830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000509903.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000510756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000511049.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 ENST00000511782.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 NM_001042440.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 NM_001190442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 NM_173060.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05006648.hg.1 TC05000469.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000309190.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000338252.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000341926.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000348386.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000395812.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000395813.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000421689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000504465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000504532.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000505143.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000506811.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508117.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508197.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508608.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508830.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000509903.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000510156.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000510756.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511049.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511097.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511782.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000512620.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000514055.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000514845.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001042440.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001190442.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001284212.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001284213.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_173060.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000309190.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000338252.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000341926.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000348386.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000395812.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000395813.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000421689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000504465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000504532.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000505143.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000506811.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508117.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508197.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508608.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000508830.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000509903.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000510156.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000510756.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511049.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511097.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000511782.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000512620.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000514055.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 ENST00000514845.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001042440.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001190442.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001284212.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_001284213.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 NM_173060.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500193584.hg.1 TC0500013204.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline