| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000481842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000481842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010127.hg.1 |
ENST00000368977.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010127.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010127.hg.1 |
TC06000866.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162181.hg.1 |
TC0600009051.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000368977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000421954.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000430458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000431865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000437715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000481842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000481842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
ENST00000486863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009051.hg.1 |
NM_145315.4 |
Refseq |