Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00990356_m1
          See other SSTR2 GE Assays ›
          Gene Symbol:
          SSTR2
          Gene Name:
          somatostatin receptor 2
          Gene Aliases:
          SST2
          Chromosome Location:
          Chr.17: 73165010 - 73176633 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00990356_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SSTR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001050.3 NP_001041.1 1 - 2 284 54
          GenBank mRNA AK290745.1 - 1 - 2 284 54
          AK313866.1 - 1 - 2 262 54
          BC019610.1 - 1 - 2 273 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012303.hg
          This assay targets the same location as HTA transcript cluster(s) TC17000821.hg

          Additional Information:

          This assay is in the 5'UTR region of the BC019610.1, NM_001050.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SSTR2

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          negative regulation of cell proliferation
          somatostatin signaling pathway
          cellular response to glucocorticoid stimulus
          cellular response to estradiol stimulus
          G-protein coupled receptor activity
          somatostatin receptor activity
          protein binding
          PDZ domain binding
          neuropeptide binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17010556.hg.1 ENST00000357585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010556.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010556.hg.1 TC17000821.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17010558.hg.1 ENST00000315332.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010558.hg.1 ENST00000357585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010558.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010558.hg.1 TC17000821.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000569655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010555.hg.1 ENST00000357585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010555.hg.1 NM_001050.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010555.hg.1 TC17000821.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203709.hg.1 TC1700012303.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203707.hg.1 TC1700012303.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000569655.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 ENST00000579323.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012303.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203712.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203712.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203712.hg.1 ENST00000357585.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700203712.hg.1 NM_001050.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700203712.hg.1 TC1700012303.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 211133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline