Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00991530_s1
          See other BIN3-IT1 GE Assays ›
          Gene Symbol:
          BIN3-IT1
          Gene Name:
          BIN3 intronic transcript 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 22640371 - 22642209 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00991530_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BIN3-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027715.1 114
          GenBank mRNA AK024169.1 - 114
          BC013100.1 - 114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY271447.1 - Chr. 8 - 22640374 - 22640833 [+] on Build GRCh38 114
          GenBankmRNA AU147638.2 - Chr. 8 - 22640371 - 22640928 [+] on Build GRCh38 114

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P72646 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P72646 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18305 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40830 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P72646 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P72646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006796 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006802 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006802 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006796 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16078351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26288852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06006780 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P72646 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006786 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006799 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006830 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006803 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006811 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16078348 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006814 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006817 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006795 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006819 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16078345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006805 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16078346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006789 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006801 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006820 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006793 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16078351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006790 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006797 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006808 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06006783 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8149719 - -
          Affymetrix Human Gene 1.1 ST Array 8149718 - -
          Affymetrix HT HG-U133+ PM Array Plate 207287_PM_at - -
          Affymetrix Human Genome U219 Array 11745305_at - -
          Affymetrix Human Genome U219 Array 11745305_PM_at - -
          Human Genome U133 Plus 2.0 Array 207287_PM_at - -
          Human Genome U133 Plus 2.0 Array 207287_at - -
          Human Genome U133A 2.0 Array 207287_at - -
          Human Genome U133A 2.0 Array 207287_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw525934 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207287_at5 - -
          Human Genome U133 Set 207287_at - -
          Human Genome U133 Set 207287_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035597 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035597 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122713 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3307196 BC013100.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307196 NR_027715.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307196 AK024169.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1811608 BC013100.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1811608 NR_027715.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811608 BC013100.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811608 AK024169.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1811608 BC013100.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1811608 NR_027715.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1811608 AK024169.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline