Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00992286_g1
          See other TRIP12 GE Assays ›
          Gene Symbol:
          TRIP12
          Gene Name:
          thyroid hormone receptor interactor 12
          Gene Aliases:
          TRIP-12, ULF
          Chromosome Location:
          Chr.2: 229763837 - 229923234 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00992286_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIP12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001284214.1 NP_001271143.1 10 - 11 1815 92
          NM_001284215.1 NP_001271144.1 8 - 9 1559 92
          NM_001284216.1 NP_001271145.1 7 - 8 780 92
          NM_004238.2 NP_004229.1 9 - 10 1671 92
          XM_005246954.4 XP_005247011.1 10 - 11 1839 92
          XM_005246955.4 XP_005247012.1 10 - 11 1838 92
          XM_005246956.4 XP_005247013.1 10 - 11 1818 92
          XM_005246957.4 XP_005247014.1 10 - 11 1805 92
          XM_005246958.4 XP_005247015.1 10 - 11 1839 92
          XM_005246960.4 XP_005247017.1 11 - 12 1752 92
          XM_005246961.3 XP_005247018.1 9 - 10 1689 92
          XM_005246962.4 XP_005247019.1 9 - 10 1689 92
          XM_006712852.3 XP_006712915.1 9 - 10 1642 92
          XM_011512180.2 XP_011510482.1 10 - 11 1792 92
          XM_011512181.2 XP_011510483.1 10 - 11 1790 92
          XM_011512183.2 XP_011510485.1 9 - 10 1646 92
          XM_017005272.1 XP_016860761.1 11 - 12 1898 92
          XM_017005273.1 XP_016860762.1 10 - 11 1765 92
          XM_017005274.1 XP_016860763.1 10 - 11 1713 92
          XM_017005275.1 XP_016860764.1 10 - 11 1771 92
          XM_017005276.1 XP_016860765.1 10 - 11 1776 92
          XM_017005277.1 XP_016860766.1 11 - 12 1916 92
          XM_017005278.1 XP_016860767.1 10 - 11 1731 92
          XM_017005279.1 XP_016860768.1 10 - 11 1803 92
          XM_017005280.1 XP_016860769.1 10 - 11 1792 92
          XM_017005281.1 XP_016860770.1 10 - 11 1792 92
          XM_017005282.1 XP_016860771.1 11 - 12 1751 92
          XM_017005283.1 XP_016860772.1 10 - 11 1839 92
          XM_017005284.1 XP_016860773.1 11 - 12 1752 92
          XM_017005285.1 XP_016860774.1 9 - 10 1676 92
          XM_017005286.1 XP_016860775.1 9 - 10 1711 92
          XM_017005287.1 XP_016860776.1 9 - 10 1692 92
          XM_017005288.1 XP_016860777.1 9 - 10 1654 92
          XM_017005289.1 XP_016860778.1 11 - 12 1752 92
          XM_017005290.1 XP_016860779.1 11 - 12 1752 92
          XM_017005291.1 XP_016860780.1 10 - 11 1602 92
          XM_017005292.1 XP_016860781.1 9 - 10 1689 92
          XM_017005293.1 XP_016860782.1 9 - 10 1689 92
          GenBank mRNA AK310971.1 - 9 - 10 1689 92
          BC113891.1 - 7 - 8 674 40083155 92
          BC114556.1 - 10 - 11 1709 40083165 92
          D28476.1 - 9 - 10 1601 92
          EU489742.1 - 8 - 9 1513 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015963.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002848.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIP12

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          DNA repair
          cellular response to DNA damage stimulus
          embryo development
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of histone H2A K63-linked ubiquitination
          negative regulation of double-strand break repair
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          thyroid hormone receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02045990.hg.1 ENST00000283943.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045990.hg.1 ENST00000389044.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045990.hg.1 ENST00000389045.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045990.hg.1 NM_004238.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02045990.hg.1 TC02002848.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000283943.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000389044.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000389045.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000479037.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284215.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284216.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_004238.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000283943.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000389044.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000389045.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 ENST00000479037.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284215.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_001284216.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 NM_004238.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217439.hg.1 TC0200015963.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline