Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00996832_m1
          See other SLC18A2 GE Assays ›
          Gene Symbol:
          SLC18A2
          Gene Name:
          solute carrier family 18 member A2
          Gene Aliases:
          SVAT, SVMT, VAT2, VMAT2
          Chromosome Location:
          Chr.10: 117241073 - 117279430 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00996832_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC18A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003054.4 NP_003045.2 12 - 13 1281 95
          GenBank mRNA AK314997.1 - 12 - 13 1234 95
          BC030593.1 - 11 - 12 1539 95
          BC108927.1 - 11 - 12 1146 40025174 95
          BC108928.2 - 11 - 12 1146 40025175 95
          L14269.1 - 12 - 13 1151 95
          L23205.1 - 12 - 13 1231 95
          X71354.1 - 12 - 13 1181 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009017.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000850.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC18A2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          serotonin transport
          chemical synaptic transmission
          neurotransmitter secretion
          aging
          locomotory behavior
          response to herbicide
          post-embryonic development
          response to zinc ion
          aminergic neurotransmitter loading into synaptic vesicle
          monoamine transport
          dopamine transport
          insulin secretion
          endocytic recycling
          cellular response to drug
          sequestering of neurotransmitter
          response to cocaine
          glucose homeostasis
          response to starvation
          response to corticosterone
          negative regulation of neurotransmitter transport
          transmembrane transport
          cellular response to ammonium ion
          neurotransmitter loading into synaptic vesicle
          negative regulation of reactive oxygen species biosynthetic process
          amine transmembrane transporter activity
          drug binding
          monoamine transmembrane transporter activity
          serotonin transmembrane transporter activity
          enzyme binding
          heat shock protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010326.hg.1 ENST00000298472.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010326.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010326.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010326.hg.1 TC10000850.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000161795.hg.1 TC1000009017.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline