| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
ENST00000536044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
ENST00000540413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
ENST00000542049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011596.hg.1 |
TC19000866.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011597.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011597.hg.1 |
ENST00000540413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011597.hg.1 |
ENST00000542049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011597.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19011597.hg.1 |
TC19000866.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900168688.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900168688.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900168688.hg.1 |
TC1900008825.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900168688.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900168688.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |