Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001352_g1
          See other CCDC27 GE Assays ›
          Gene Symbol:
          CCDC27
          Gene Name:
          coiled-coil domain containing 27
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 3752398 - 3771645 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001352_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCDC27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152492.2 NP_689705.2 6 - 7 1033 71
          XM_011540780.1 XP_011539082.1 6 - 7 1036 71
          XM_017000402.1 XP_016855891.1 6 - 7 1036 71
          XM_017000403.1 XP_016855892.1 6 - 7 1036 71
          XM_017000404.1 XP_016855893.1 6 - 7 1036 71
          XM_017000405.1 XP_016855894.1 6 - 7 1036 71
          XM_017000406.1 XP_016855895.1 7 - 8 850 71
          GenBank mRNA AK057387.1 - 6 - 7 1036 71
          BC112108.1 - 6 - 7 1034 8327576 71
          BC112112.1 - 6 - 7 1033 8327580 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006623.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000072.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCDC27

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01001160.hg.1 ENST00000294600.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01001160.hg.1 ENST00000462521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01001160.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01001160.hg.1 TC01000072.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100145826.hg.1 TC0100006623.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000294600.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 ENST00000462521.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006623.hg.1 NM_152492.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline