| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022610.hg.1 |
ENST00000287748.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022610.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022610.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022610.hg.1 |
TC03001319.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022607.hg.1 |
ENST00000287748.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022607.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022607.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022607.hg.1 |
TC03001319.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178918.hg.1 |
TC0300010841.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000441172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
ENST00000470991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010841.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
ENST00000287748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
NM_001304386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
NM_144634.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178920.hg.1 |
TC0300010841.hg.1 |
Clariom D TC |