Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01006331_g1
          See other GLP1R GE Assays ›
          Gene Symbol:
          GLP1R
          Gene Name:
          glucagon like peptide 1 receptor
          Gene Aliases:
          GLP-1, GLP-1-R, GLP-1R
          Chromosome Location:
          Chr.6: 39048781 - 39091303 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01006331_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GLP1R
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002062.4 NP_002053.3 6 - 7 721 117
          NR_136562.1 6 - 7 721 117
          NR_136563.1 6 - 7 721 117
          XM_017010750.1 XP_016866239.1 5 - 6 1256 117
          XM_017010751.1 XP_016866240.1 5 - 6 1259 117
          GenBank mRNA BC112126.1 - 6 - 7 733 8327594 117
          BC113493.1 - 6 - 7 721 8322545 117
          KR138540.1 - 6 - 7 687 117
          L23503.1 - 6 - 7 686 117
          U01104.1 - 6 - 7 703 117
          U01156.1 - 6 - 7 661 117
          U01157.1 - 6 - 7 661 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007946.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000557.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GLP1R

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to stress
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          learning or memory
          regulation of heart contraction
          cAMP-mediated signaling
          positive regulation of blood pressure
          regulation of insulin secretion
          cellular response to glucagon stimulus
          transmembrane signaling receptor activity
          glucagon receptor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06006188.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006188.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006188.hg.1 TC06000557.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007946.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600155042.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600155042.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600155042.hg.1 ENST00000373256.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600155042.hg.1 NM_002062.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600155042.hg.1 TC0600007946.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline