Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01007070_m1
          See other CASZ1 GE Assays ›
          Gene Symbol:
          CASZ1
          Gene Name:
          castor zinc finger 1
          Gene Aliases:
          CAS11, CST, SRG, ZNF693, dJ734G22.1
          Chromosome Location:
          Chr.1: 10636602 - 10796676 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01007070_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CASZ1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079843.2 NP_001073312.1 13 - 14 3228 93
          NM_017766.4 NP_060236.3 13 - 14 3228 93
          XM_005263479.3 XP_005263536.1 11 - 12 3092 93
          XM_011541635.2 XP_011539937.1 11 - 12 3091 93
          XM_017001539.1 XP_016857028.1 11 - 12 3092 93
          XM_017001540.1 XP_016857029.1 13 - 14 3202 93
          XM_017001541.1 XP_016857030.1 13 - 14 3264 93
          GenBank mRNA AK000328.1 - 9 - 10 2542 93
          AK092289.1 - 13 - 14 3202 93
          AK225324.1 - 9 - 10 2542 93
          BC004410.1 - 4 - 5 1227 93
          BC051883.2 - 10 - 11 2898 5534858 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012849.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002191.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CASZ1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          central nervous system development
          regulation of neuron differentiation
          DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01033750.hg.1 ENST00000344008.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033750.hg.1 ENST00000377022.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033750.hg.1 NM_001079843.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033750.hg.1 NM_017766.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033750.hg.1 TC01002191.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 ENST00000344008.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 ENST00000377022.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 NM_001079843.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 NM_017766.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 ENST00000344008.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 ENST00000377022.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 NM_001079843.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 NM_017766.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194873.hg.1 TC0100012849.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline