Search
Search

PICALM| Gene Symbol: PICALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005274323.2 | XP_005274380.1 | 14 - 15 | 1815 | 96 | |
| XM_005274325.2 | XP_005274382.1 | 14 - 15 | 1815 | 96 | ||
| XM_005274328.2 | XP_005274385.1 | 14 - 15 | 1815 | 96 | ||
| XM_005274329.3 | XP_005274386.1 | 14 - 15 | 1815 | 96 | ||
| XM_005274332.2 | XP_005274389.1 | 13 - 14 | 1665 | 96 | ||
| XM_005274334.2 | XP_005274391.1 | 14 - 15 | 1815 | 96 | ||
| XM_005274335.2 | XP_005274392.1 | 13 - 14 | 1665 | 96 | ||
| XM_005274336.2 | XP_005274393.1 | 13 - 14 | 1665 | 96 | ||
| XM_006718700.2 | XP_006718763.1 | 14 - 15 | 1794 | 96 | ||
| XM_017018381.1 | XP_016873870.1 | 14 - 15 | 1794 | 96 | ||
| XM_017018384.1 | XP_016873873.1 | 13 - 14 | 1665 | 96 | ||
| XM_017018385.1 | XP_016873874.1 | 14 - 15 | 1815 | 96 | ||
| XM_017018387.1 | XP_016873876.1 | 13 - 14 | 1665 | 96 | ||
| GenBank mRNA | AK297062.1 | - | 5 - 6 | 566 | 96 | |
Set Membership: |
|