Search
Search
| Gene Symbol: EBF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005463.2 | NP_001005463.1 | 15 - 16 | 1679 | 73 | |
| XM_005252668.3 | XP_005252725.1 | 15 - 16 | 2261 | 73 | ||
| XM_005252669.3 | XP_005252726.1 | 15 - 16 | 2234 | 73 | ||
| XM_006717743.3 | XP_006717806.1 | 15 - 16 | 2153 | 73 | ||
| XM_017016027.1 | XP_016871516.1 | 14 - 15 | 1964 | 73 | ||
| GenBank mRNA | AL832828.1 | - | 15 - 16 | 1606 | 73 | |
| BC126130.1 | - | 15 - 16 | 1670 | 8991846 | 73 | |
| BC130479.1 | - | 15 - 16 | 1670 | 40146510 | 73 | |
Set Membership: |
|