Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01011247_m1
          See other WNT1 GE Assays ›
          Gene Symbol:
          WNT1
          Gene Name:
          Wnt family member 1
          Gene Aliases:
          BMND16, INT1, OI15
          Chromosome Location:
          Chr.12: 48978322 - 48982620 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01011247_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WNT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005430.4 NP_005421.1 1 - 2 434 108
          GenBank mRNA BC074798.2 - 1 - 2 141 108
          BC074799.2 - 1 - 2 141 108
          BT019429.1 - 1 - 2 105 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007538.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000360.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WNT1

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          branching involved in ureteric bud morphogenesis
          cell-cell signaling
          myoblast fusion
          cell proliferation
          positive regulation of cell proliferation
          response to wounding
          positive regulation of lamellipodium assembly
          negative regulation of cell-substrate adhesion
          myotube differentiation
          Wnt signaling pathway
          spinal cord association neuron differentiation
          diencephalon development
          cerebellum development
          central nervous system morphogenesis
          cerebellum formation
          forebrain anterior/posterior pattern specification
          midbrain-hindbrain boundary maturation during brain development
          neurogenesis
          metencephalon development
          negative regulation of cell-cell adhesion
          neuron differentiation
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          midbrain development
          midbrain-hindbrain boundary development
          organ regeneration
          T cell differentiation in thymus
          cell proliferation in midbrain
          astrocyte-dopaminergic neuron signaling
          inner ear morphogenesis
          signal transduction in response to DNA damage
          negative regulation of apoptotic process
          positive regulation of insulin-like growth factor receptor signaling pathway
          cell fate commitment
          fat cell differentiation
          negative regulation of cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          neuron fate commitment
          neuron fate determination
          Spemann organizer formation
          canonical Wnt signaling pathway
          bone development
          positive regulation of dermatome development
          hepatocyte differentiation
          cellular response to peptide hormone stimulus
          hematopoietic stem cell proliferation
          dopaminergic neuron differentiation
          positive regulation of hematopoietic stem cell proliferation
          negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          midbrain dopaminergic neuron differentiation
          embryonic brain development
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of cellular senescence
          receptor binding
          frizzled binding
          cytokine activity
          morphogen activity
          protein domain specific binding
          receptor agonist activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12004468.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12004468.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004468.hg.1 TC12000360.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004467.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004467.hg.1 TC12000360.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12004469.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12004469.hg.1 NM_005430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004469.hg.1 TC12000360.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153015.hg.1 TC1200007538.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153014.hg.1 TC1200007538.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153013.hg.1 TC1200007538.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000293549.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007538.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153012.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153012.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153012.hg.1 ENST00000613114.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200153012.hg.1 NM_005430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200153012.hg.1 TC1200007538.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline