Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01012666_m1
          See other THAP3 GE Assays ›
          Gene Symbol:
          THAP3
          Gene Name:
          THAP domain containing 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 6624868 - 6635595 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01012666_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: THAP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195752.2 NP_001182681.1 2 - 3 232 83
          NM_001195753.2 NP_001182682.1 2 - 3 232 83
          NM_001394496.1 NP_001381425.1 1 - 2 112 83
          NM_001394497.1 NP_001381426.1 1 - 2 112 83
          NM_001394498.1 NP_001381427.1 1 - 2 112 83
          NM_001394499.1 NP_001381428.1 1 - 2 112 83
          NM_001394500.1 NP_001381429.1 1 - 2 112 83
          NM_138350.4 NP_612359.2 1 - 2 112 83
          XM_024450686.2 XP_024306454.1 2 - 3 232 83
          XM_047433628.1 XP_047289584.1 2 - 3 232 83
          GenBank mRNA AK096217.1 - 1 - 2 103 83
          BC092427.1 - 2 - 3 235 83
          BE905510.1 - 204 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006699.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000093.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene THAP3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription from RNA polymerase II promoter
          DNA binding
          protein binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 ENST00000401022.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 NR_033990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 NR_033990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018250.hg.1 NR_033990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000054650.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000307896.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000377627.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000472925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000480647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000480647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000480647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484669.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484676.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484676.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000484676.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 ENST00000487819.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195752.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_001195753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006699.hg.1 NM_138350.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 ENST00000401022.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 ENST00000418362.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 NR_033990.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 NR_033990.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018250.hg.1 NR_033990.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline