| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8052439 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8052442 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8052441 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8052440 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8052438 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
227940_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
227941_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030116.hg.1 |
ENST00000420918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030116.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030116.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030117.hg.1 |
ENST00000420918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030117.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030117.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030110.hg.1 |
ENST00000420918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030110.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030110.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030112.hg.1 |
ENST00000420918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030112.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030112.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030108.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030108.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030109.hg.1 |
ENST00000420918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030109.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030109.hg.1 |
TC02001890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192776.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012731.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192787.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192779.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192774.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
ENST00000542553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192775.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192777.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000417691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000420918.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000422740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
ENST00000443946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
NR_036496.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192786.hg.1 |
TC0200012731.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
227940_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
227941_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
227941_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
227940_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227941_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
227940_at10 |
- |
- |
| Human Genome U95 Set |
51820_PM_at |
- |
- |
| Human Genome U95 Set |
51820_at |
- |
- |
| Human Genome U95 Set |
73714_PM_at |
- |
- |
| Human Genome U95 Set |
73714_at |
- |
- |
| Human Genome U95 Set |
90198_PM_at |
- |
- |
| Human Genome U95 Set |
90198_at |
- |
- |
| Human Genome U133 Set |
227940_PM_at |
- |
- |
| Human Genome U133 Set |
227941_PM_at |
- |
- |
| Human Genome U133 Set |
227940_at |
- |
- |
| Human Genome U133 Set |
227941_at |
- |
- |