Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01015824_m1
          See other CA8 GE Assays ›
          Gene Symbol:
          CA8
          Gene Name:
          carbonic anhydrase 8
          Gene Aliases:
          CA-RP, CA-VIII, CALS, CAMRQ3, CARP
          Chromosome Location:
          Chr.8: 60185420 - 60281423 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01015824_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CA8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321837.1 NP_001308766.1 2 - 3 569 156
          NM_001321838.1 NP_001308767.1 2 - 3 569 156
          NM_001321839.1 NP_001308768.1 2 - 3 569 156
          NM_004056.5 NP_004047.3 2 - 3 569 156
          NR_135821.1 2 - 3 569 156
          XM_011517587.2 XP_011515889.1 2 - 3 541 156
          XM_011517588.2 XP_011515890.1 2 - 3 541 156
          GenBank mRNA AK090655.1 - 2 - 3 541 156
          AK289470.1 - 2 - 3 541 156
          AK293999.1 - 1 - 2 160 156
          AK314538.1 - 2 - 3 542 156
          AY075022.1 - 2 - 3 546 156
          BC069744.1 - 2 - 3 538 7262540 156
          BC069794.1 - 2 - 3 573 7262552 156
          BC108929.2 - 2 - 3 494 40025442 156
          BM462961.1 - 2 - 3 542 156
          L04656.1 - 2 - 3 291 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800010575.hg
          This assay targets the same location as HTA transcript cluster(s) TC08001253.hg

          Additional Information:

          This assay is in the 5'UTR region of the AK293999.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CA8

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          phosphatidylinositol-mediated signaling
          carbonate dehydratase activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08016205.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08016205.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08016205.hg.1 TC08001253.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08016203.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08016203.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08016203.hg.1 TC08001253.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08016204.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08016204.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08016204.hg.1 TC08001253.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08016206.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08016206.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08016206.hg.1 TC08001253.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000528666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay TC0800010575.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171843.hg.1 TC0800010575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171847.hg.1 TC0800010575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171844.hg.1 TC0800010575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171845.hg.1 TC0800010575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000317995.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000524872.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 ENST00000529918.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 NM_004056.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800171842.hg.1 TC0800010575.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 159072 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline