Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01016888_m1
See other CALCR GE Assays ›
Gene Symbol:
CALCR
Gene Name:
calcitonin receptor
Gene Aliases:
CRT, CT-R, CTR, CTR1
Chromosome Location:
Chr.7: 93424487 - 93574730 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01016888_m1
Size
FAM (900 nM Primer) | XS: 75 rxns
FAM (900 nM Primer) | S: 360 rxns
FAM (900 nM Primer) | M: 750 rxns
FAM (900 nM Primer) | L: 2,900 rxns
FAM PL (350 nM Primer Limited) | S: 360 rxns
FAM PL (350 nM Primer Limited) | M: 750 rxns
FAM PL (350 nM Primer Limited) | L: 2,900 rxns
VIC (900 nM Primer) | S: 360 rxns
VIC (900 nM Primer) | M: 750 rxns
VIC (900 nM Primer) | L: 2,900 rxns
VIC PL (150 nM Primer Limited) | S: 360 rxns
VIC PL (150 nM Primer Limited) | M: 750 rxns
VIC PL (150 nM Primer Limited) | L: 2,900 rxns
ABY PL (350 nM Primer Limited) | S: 360 rxns
ABY PL (350 nM Primer Limited) | M: 750 rxns
ABY PL (350 nM Primer Limited) | L: 2,900 rxns
JUN PL (350 nM Primer Limited) | S: 360 rxns
JUN PL (350 nM Primer Limited) | M: 750 rxns
JUN PL (350 nM Primer Limited) | L: 2,900 rxns
CYN5 PL (350 nM Primer Limited) | S: 360 rxns
CYN5 PL (350 nM Primer Limited) | M: 750 rxns
CYN5 PL (350 nM Primer Limited) | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CALCR
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
Amplicon Length
RefSeq
NM_001164737.1
NP_001158209.1
9 - 10
927
156
GenBank mRNA
AJ831407.1
-
6 - 7
573
156
L00587.1
-
8 - 9
820
156
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
G-protein coupled receptor
Homo sapiens
Probe spans exons
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
positive regulation of adenylate cyclase activity
response to glucocorticoid
protein localization to plasma membrane
receptor activity
calcitonin receptor activity
protein binding
protein transporter activity
calcitonin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.60.1-2026-secondary-format-patch