| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
ENST00000263334.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
ENST00000263335.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
ENST00000348715.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
ENST00000429538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
ENST00000497038.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
NM_003466.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
NM_013952.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
NM_013953.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036093.hg.1 |
TC02002222.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000263334.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000263335.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000348715.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000429538.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000465084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000468980.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000485840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000497038.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_003466.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_013952.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_013953.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000263334.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000263335.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000348715.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000429538.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000465084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000468980.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000485840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
ENST00000497038.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_003466.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_013952.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
NM_013953.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200202145.hg.1 |
TC0200013925.hg.1 |
Clariom D TC |