Search
Search

LLGL1| Gene Symbol: LLGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523851.3 | XP_011522153.1 | 15 - 16 | 2101 | 136 | |
| XM_011523856.2 | XP_011522158.2 | 15 - 16 | 2101 | 136 | ||
| XM_047436003.1 | XP_047291959.1 | 15 - 16 | 2101 | 136 | ||
| XM_047436005.1 | XP_047291961.1 | 15 - 16 | 2098 | 136 | ||
| XM_047436006.1 | XP_047291962.1 | 15 - 16 | 2101 | 136 | ||
| XM_047436007.1 | XP_047291963.1 | 15 - 16 | 2098 | 136 | ||
| XM_047436008.1 | XP_047291964.1 | 15 - 16 | 2101 | 136 | ||
Set Membership: |
|