| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012509.hg.1 |
ENST00000313135.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012509.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012509.hg.1 |
TC17000950.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700170259.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170259.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170259.hg.1 |
TC1700009249.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700170259.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170259.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170260.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170260.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170260.hg.1 |
TC1700009249.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700170260.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170260.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
ENST00000313135.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009249.hg.1 |
NM_018949.2 |
Refseq |