| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018763.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018763.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018763.hg.1 |
TC08001444.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800176029.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800176029.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800176029.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800176029.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800176029.hg.1 |
TC0800011158.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
ENST00000286688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011158.hg.1 |
NM_177965.3 |
Refseq |