Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01031724_m1
          See other VIL1 GE Assays ›
          Gene Symbol:
          VIL1
          Gene Name:
          villin 1
          Gene Aliases:
          D2S1471, VIL
          Chromosome Location:
          Chr.2: 218419115 - 218449525 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01031724_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VIL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007127.2 NP_009058.2 13 - 14 1564 84
          GenBank mRNA AK223398.1 - 13 - 14 1558 84
          AK300965.1 - 8 - 9 795 84
          AK313709.1 - 13 - 14 1556 84
          X12901.1 - 12 - 13 1523 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010800.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001303.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene VIL1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intestinal D-glucose absorption
          protein complex assembly
          apoptotic process
          epidermal growth factor receptor signaling pathway
          regulation of cell shape
          response to bacterium
          positive regulation of epithelial cell migration
          actin filament polymerization
          actin filament depolymerization
          positive regulation of cell migration
          positive regulation of actin filament depolymerization
          epithelial cell differentiation
          positive regulation of actin filament bundle assembly
          regulation of microvillus length
          cellular response to hepatocyte growth factor stimulus
          positive regulation of multicellular organism growth
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          actin nucleation
          actin filament severing
          barbed-end actin filament capping
          regulation of actin nucleation
          actin filament capping
          cytoplasmic actin-based contraction involved in cell motility
          regulation of wound healing
          cellular response to epidermal growth factor stimulus
          positive regulation of establishment of protein localization to plasma membrane
          terminal web assembly
          regulation of lamellipodium morphogenesis
          positive regulation of lamellipodium morphogenesis
          calcium ion binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          lysophosphatidic acid binding
          identical protein binding
          protein homodimerization activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          actin filament binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000248444.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000392114.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000419986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000419986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000419986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000419986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000419986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000440053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 ENST00000454069.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010800.hg.1 NM_007127.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 191456 - -
          ABI Human Genome Survey Microarray Version 2.0 234681 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline