Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01038439_m1
          See other RARS2 GE Assays ›
          Gene Symbol:
          RARS2
          Gene Name:
          arginyl-tRNA synthetase 2, mitochondrial
          Gene Aliases:
          ArgRS, DALRD2, PCH6, PRO1992, RARSL
          Chromosome Location:
          Chr.6: 87513744 - 87590032 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01038439_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RARS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318785.1 NP_001305714.1 10 - 11 1212 160
          NM_020320.4 NP_064716.2 11 - 12 1047 160
          NR_134857.1 11 - 12 1047 156
          XM_011535948.2 XP_011534250.1 11 - 12 1032 160
          XM_011535949.2 XP_011534251.1 11 - 12 1032 160
          XM_017011072.1 XP_016866561.1 11 - 12 1213 160
          XM_017011073.1 XP_016866562.1 12 - 13 1431 160
          XM_017011074.1 XP_016866563.1 11 - 12 1034 160
          XM_017011075.1 XP_016866564.1 10 - 11 1157 160
          XM_017011076.1 XP_016866565.1 12 - 13 1375 160
          XM_017011077.1 XP_016866566.1 10 - 11 934 160
          XM_017011078.1 XP_016866567.1 10 - 11 978 160
          XM_017011079.1 XP_016866568.1 11 - 12 1213 160
          XM_017011080.1 XP_016866569.1 12 - 13 1375 160
          XM_017011081.1 XP_016866570.1 10 - 11 978 160
          GenBank mRNA AK023550.1 - 11 - 12 990 160
          AK093934.1 - 11 - 12 1420 160
          AK315669.1 - 11 - 12 984 160
          BC010420.1 - 11 - 12 1018 3613406 160
          BC105784.1 - 10 - 11 1151 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012512.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001928.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RARS2

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA aminoacylation for protein translation
          arginyl-tRNA aminoacylation
          mitochondrial translation
          arginine-tRNA ligase activity
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06023625.hg.1 ENST00000369523.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023625.hg.1 ENST00000369536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023625.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06023625.hg.1 TC06001928.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186191.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600186191.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186191.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600186191.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186191.hg.1 TC0600012512.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline