| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003673.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003673.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003673.hg.1 |
TC01000231.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100149566.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149566.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100149566.hg.1 |
ENST00000375460.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149566.hg.1 |
NM_016233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100149566.hg.1 |
TC0100007103.hg.1 |
Clariom D TC |