| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000356245.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000394123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000507878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000517947.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000518726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000518726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000518726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000519832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520006.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520177.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000520578.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000522761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000523519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
ENST00000627077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_005754.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009131.hg.1 |
NM_198395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007233.hg.1 |
ENST00000445475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007233.hg.1 |
ENST00000445475.1 |
Ensembl |