Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01046739_m1
          See other PDCD10 GE Assays ›
          Gene Symbol:
          PDCD10
          Gene Name:
          programmed cell death 10
          Gene Aliases:
          CCM3, TFAR15
          Chromosome Location:
          Chr.3: 167683891 - 167735690 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01046739_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDCD10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145859.1 NP_665858.1 1 - 2 59 88
          GenBank mRNA BC016353.1 - 1 - 2 63 4092244 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300013051.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001976.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC016353.1,NM_145859.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDCD10

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of cell migration
          positive regulation of stress-activated MAPK cascade
          positive regulation of peptidyl-serine phosphorylation
          regulation of Rho protein signal transduction
          intrinsic apoptotic signaling pathway in response to hydrogen peroxide
          response to hydrogen peroxide
          negative regulation of apoptotic process
          stress fiber assembly
          positive regulation of MAP kinase activity
          wound healing, spreading of cells
          positive regulation of Notch signaling pathway
          protein stabilization
          establishment of Golgi localization
          positive regulation of protein serine/threonine kinase activity
          negative regulation of cell migration involved in sprouting angiogenesis
          Golgi reassembly
          negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          protein binding
          protein kinase binding
          protein homodimerization activity
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03035365.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035365.hg.1 ENST00000487678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035365.hg.1 ENST00000492396.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035365.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035365.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 ENST00000470131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 ENST00000471885.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 ENST00000497056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 NM_145860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035390.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 ENST00000470131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 ENST00000471885.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 ENST00000497056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 NM_145860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035391.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03035393.hg.1 ENST00000470131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035393.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035393.hg.1 ENST00000497056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035393.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035393.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 ENST00000470131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 ENST00000471885.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 ENST00000497056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035394.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 ENST00000470131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 ENST00000471885.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 ENST00000473645.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 ENST00000497056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 NM_145859.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 NM_145860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03035392.hg.1 TC03001976.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000462725.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000464360.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000473645.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000475915.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000487678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000492396.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000494502.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 NM_145859.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000462725.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000464360.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000473645.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000475915.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000487678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000492396.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 ENST00000494502.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 NM_145859.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300197588.hg.1 TC0300013051.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline