| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000354546.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000356496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000377751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000521512.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000523714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
ENST00000540153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
NM_001155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
NM_001193544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027078.hg.1 |
TC05001947.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000354546.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000377751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000521512.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000523714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
NM_001155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
NM_001193544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000354546.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000377751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000521512.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
ENST00000523714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
NM_001155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
NM_001193544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187177.hg.1 |
TC0500012498.hg.1 |
Clariom D TC |