Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01050416_m1
          See other MEF2A GE Assays ›
          Gene Symbol:
          MEF2A
          Gene Name:
          myocyte enhancer factor 2A
          Gene Aliases:
          ADCAD1, RSRFC4, RSRFC9, mef2
          Chromosome Location:
          Chr.15: 99565455 - 99716488 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01050416_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MEF2A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001319206.1 NP_001306135.1 2 - 3 250 123
          NM_005587.3 NP_005578.2 2 - 3 250 123
          XM_005254914.2 XP_005254971.1 2 - 3 215 123
          XM_005254915.2 XP_005254972.1 2 - 3 478 123
          XM_005254916.3 XP_005254973.1 2 - 3 215 123
          XM_011521571.2 XP_011519873.1 2 - 3 215 123
          XM_011521581.2 XP_011519883.1 2 - 3 478 123
          XM_011521585.2 XP_011519887.1 2 - 3 478 123
          XM_011521586.2 XP_011519888.1 2 - 3 478 123
          XM_011521587.2 XP_011519889.1 2 - 3 478 123
          XM_011521590.2 XP_011519892.1 2 - 3 215 123
          XM_017022195.1 XP_016877684.1 2 - 3 165 123
          XM_017022196.1 XP_016877685.1 2 - 3 478 123
          XM_017022198.1 XP_016877687.1 2 - 3 478 123
          XM_017022200.1 XP_016877689.1 2 - 3 478 123
          XM_017022202.1 XP_016877691.1 2 - 3 478 123
          XM_017022203.1 XP_016877692.1 2 - 3 478 123
          XM_017022204.1 XP_016877693.1 2 - 3 478 123
          GenBank mRNA AL831995.1 - 2 - 3 111 123
          BC013437.2 - 2 - 3 168 3896491 123
          DA778137.1 - 2 - 3 250 123
          X68504.1 - 2 - 3 126 123

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC013437.2, NM_001319206.1, NM_005587.3, XM_005254914.2, XM_005254915.2, XM_005254916.3, XM_011521571.2, XM_011521581.2, XM_011521585.2, XM_011521586.2, XM_011521587.2, XM_011521590.2, XM_017022195.1, XM_017022196.1, XM_017022198.1, XM_017022200.1, XM_017022202.1, XM_017022203.1, XM_017022204.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MEF2A

          Panther Classification:

          Molecular Function -

          MADS box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          heart development
          muscle organ development
          positive regulation of cardiac muscle hypertrophy
          positive regulation of transcription from RNA polymerase II promoter
          mitochondrion distribution
          dendrite morphogenesis
          positive regulation of muscle cell differentiation
          ventricular cardiac myofibril assembly
          cardiac conduction
          ERK5 cascade
          cellular response to calcium ion
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II transcription coactivator activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          protein kinase binding
          activating transcription factor binding
          histone acetyltransferase binding
          histone deacetylase binding
          sequence-specific DNA binding
          SMAD binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016554.hg.1 ENST00000454867.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016554.hg.1 ENST00000411857.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016554.hg.1 ENST00000454867.2 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 191872 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline