Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01052262_m1
          See other ABI2 GE Assays ›
          Gene Symbol:
          ABI2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
          Chromosome Location:
          Chr.2: 203327874 - 203432173 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01052262_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ABI2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005759.5 NP_005750.4 8 - 9 1346 87
          XM_005246217.2 XP_005246274.1 10 - 11 1912 87
          XM_005246218.2 XP_005246275.1 9 - 10 1894 87
          XM_005246221.2 XP_005246278.1 8 - 9 1747 87
          XM_006712174.2 XP_006712237.1 10 - 11 1912 87
          XM_006712177.2 XP_006712240.1 11 - 12 1988 87
          XM_006712179.2 XP_006712242.1 9 - 10 1765 87
          XM_006712180.2 XP_006712243.1 9 - 10 1729 87
          XM_011510472.2 XP_011508774.1 9 - 10 1762 87
          XM_011510473.2 XP_011508775.1 9 - 10 1211 87
          XM_017003118.1 XP_016858607.1 10 - 11 1909 87
          XM_017003120.1 XP_016858609.1 10 - 11 1909 87
          XM_017003123.1 XP_016858612.1 8 - 9 1744 87
          XM_017003124.1 XP_016858613.1 8 - 9 1711 87
          XM_017003125.1 XP_016858614.1 8 - 9 1708 87
          XM_017003127.1 XP_016858616.1 8 - 9 1708 87
          XM_017003133.1 XP_016858622.1 10 - 11 1841 87
          XM_017003137.1 XP_016858626.1 10 - 11 1802 87
          XM_017003138.1 XP_016858627.1 8 - 9 1579 87
          XM_017003139.1 XP_016858628.1 9 - 10 1787 87
          XM_017003140.1 XP_016858629.1 9 - 10 1784 87
          XM_017003141.1 XP_016858630.1 7 - 8 1564 87
          XM_017003142.1 XP_016858631.1 7 - 8 1561 87
          GenBank mRNA AK299672.1 - 8 - 9 837 87
          AK299688.1 - 5 - 6 638 87
          AK299750.1 - 3 - 4 284 87
          AK316343.1 - 3 - 4 338 87
          BC001439.2 - 8 - 9 1095 3138341 87
          BT009920.1 - 8 - 9 1070 87
          U31089.1 - 7 - 8 815 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC02001198.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ABI2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cytoskeleton organization
          actin polymerization or depolymerization
          viral process
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          positive regulation of Arp2/3 complex-mediated actin nucleation
          DNA binding
          protein binding
          cytoskeletal adaptor activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          Rac GTPase binding
          proline-rich region binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000261016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000261017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000261018.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000295851.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000422511.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000424558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 ENST00000430418.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 NM_005759.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018428.hg.1 TC02001198.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000261016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000261017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000261018.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000295851.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000422511.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000424558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000430418.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 ENST00000430574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 NM_005759.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018426.hg.1 TC02001198.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline