Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01052828_m1
          See other TOP1 GE Assays ›
          Gene Symbol:
          TOP1
          Gene Name:
          topoisomerase (DNA) I
          Gene Aliases:
          TOPI
          Chromosome Location:
          Chr.20: 41028822 - 41124487 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01052828_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TOP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003286.2 NP_003277.1 5 - 6 582 152
          GenBank mRNA AK225095.1 - 5 - 6 581 152
          AK292943.1 - 5 - 6 581 152
          AK310508.1 - 4 - 5 464 152
          BC004475.2 - 5 - 6 564 152
          BC067349.1 - 5 - 6 561 152
          BC136297.1 - 5 - 6 433 9020284 152
          J03250.1 - 5 - 6 547 152
          U07804.1 - 5 - 6 326 152
          U07806.1 - 5 - 6 326 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007380.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000302.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TOP1

          Panther Classification:

          Molecular Function -

          DNA topoisomerase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA topological change
          chromatin remodeling
          chromosome segregation
          circadian rhythm
          programmed cell death
          viral process
          phosphorylation
          protein sumoylation
          circadian regulation of gene expression
          embryonic cleavage
          response to drug
          core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          DNA topoisomerase type I activity
          DNA topoisomerase type II (ATP-hydrolyzing) activity
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004278.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004278.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004278.hg.1 TC20000302.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20004279.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004279.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004279.hg.1 TC20000302.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20004277.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004277.hg.1 NM_003286.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004277.hg.1 TC20000302.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000151031.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151031.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000151031.hg.1 TC2000007380.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000151031.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151031.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay TC2000007380.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000151032.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151032.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000151032.hg.1 TC2000007380.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000151032.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151032.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000151033.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151033.hg.1 NM_003286.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000151033.hg.1 TC2000007380.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000151033.hg.1 ENST00000361337.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000151033.hg.1 NM_003286.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline