Search
Search
| Gene Symbol: MYOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013451.3 | NP_038479.1 | 15 - 16 | 1456 | 132 | |
| NM_133337.2 | NP_579899.1 | 15 - 16 | 1456 | 132 | ||
| XM_005269693.4 | XP_005269750.1 | 15 - 16 | 1635 | 132 | ||
| XM_005269694.4 | XP_005269751.1 | 15 - 16 | 1635 | 132 | ||
| XM_011539632.2 | XP_011537934.1 | 16 - 17 | 1829 | 132 | ||
| XM_017016068.1 | XP_016871557.1 | 14 - 15 | 1216 | 132 | ||
| XM_017016069.1 | XP_016871558.1 | 14 - 15 | 1261 | 132 | ||
| XM_017016070.1 | XP_016871559.1 | 15 - 16 | 1643 | 132 | ||
| GenBank mRNA | AB033033.2 | - | 15 - 16 | 1333 | 132 | |
| AF182316.1 | - | 15 - 16 | 1429 | 132 | ||
| AF207990.1 | - | 15 - 16 | 1333 | 132 | ||
| BC052617.1 | - | 15 - 16 | 1421 | 6422729 | 132 | |
Set Membership: |
|