Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01055462_g1
          See other BRAF GE Assays ›
          Gene Symbol:
          BRAF
          Gene Name:
          B-Raf proto-oncogene, serine/threonine kinase
          Gene Aliases:
          B-RAF1, B-raf, BRAF1, NS7, RAFB1
          Chromosome Location:
          Chr.7: 140719331 - 140924764 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01055462_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BRAF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004333.4 NP_004324.2 2 - 3 299 88
          XM_005250045.2 XP_005250102.1 2 - 3 246 88
          XM_017012558.1 XP_016868047.1 2 - 3 246 88
          XM_017012559.1 XP_016868048.1 2 - 3 246 88
          GenBank mRNA AI203435.1 - 2 - 3 271 88
          AK314346.1 - 2 - 3 246 88
          BC101757.1 - 2 - 3 260 8069263 88
          BC112079.1 - 2 - 3 260 8327547 88
          DN996440.1 - 2 - 3 497 88
          M95712.2 - 2 - 3 299 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013629.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001927.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BRAF

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          myeloid progenitor cell differentiation
          protein phosphorylation
          visual learning
          organ morphogenesis
          positive regulation of gene expression
          negative regulation of fibroblast migration
          glucose transport
          thyroid gland development
          positive regulation of peptidyl-serine phosphorylation
          somatic stem cell population maintenance
          cellular response to drug
          regulation of cell proliferation
          negative regulation of apoptotic process
          CD4-positive, alpha-beta T cell differentiation
          positive T cell selection
          response to peptide hormone
          negative regulation of neuron apoptotic process
          thymus development
          positive regulation of axon regeneration
          positive regulation of axonogenesis
          protein heterooligomerization
          positive regulation of stress fiber assembly
          response to cAMP
          long-term synaptic potentiation
          head morphogenesis
          face development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          establishment of protein localization to membrane
          positive regulation of substrate adhesion-dependent cell spreading
          negative regulation of synaptic vesicle exocytosis
          negative regulation of endothelial cell apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          calcium ion binding
          protein binding
          ATP binding
          small GTPase binding
          mitogen-activated protein kinase kinase binding
          identical protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07029959.hg.1 ENST00000288602.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029959.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07029959.hg.1 TC07001927.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000479537.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay TC0700013629.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000288602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000469930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 ENST00000497784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 NM_004333.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700202231.hg.1 TC0700013629.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline