Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01055908_m1
          See other GRHL2 GE Assays ›
          Gene Symbol:
          GRHL2
          Gene Name:
          grainyhead like transcription factor 2
          Gene Aliases:
          BOM, DFNA28, ECTDS, TFCP2L3
          Chromosome Location:
          Chr.8: 101492440 - 101678105 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01055908_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GRHL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024915.3 NP_079191.2 7 - 8 1336 102
          XM_011517305.2 XP_011515607.1 7 - 8 1367 102
          XM_011517306.2 XP_011515608.1 7 - 8 1104 102
          XM_011517307.2 XP_011515609.1 7 - 8 1334 102
          GenBank mRNA AK023844.1 - 7 - 8 1018 102
          AK292853.1 - 7 - 8 1018 102
          BC069618.1 - 7 - 8 1018 7262389 102
          BC069633.1 - 7 - 8 1078 7262401 102
          BC069638.1 - 7 - 8 1018 7262413 102
          BC129822.1 - 7 - 8 1018 40113571 102
          BC129823.1 - 7 - 8 1018 40113576 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800008416.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000629.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GRHL2

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          cardiac ventricle morphogenesis
          epithelial cell morphogenesis
          transcription from RNA polymerase II promoter
          cell adhesion
          brain development
          cell proliferation
          epidermis development
          neural tube development
          cell junction assembly
          multicellular organism growth
          embryonic digit morphogenesis
          camera-type eye development
          regulation of DNA methylation
          negative regulation of keratinocyte differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic cranial skeleton morphogenesis
          positive regulation of telomerase activity
          face development
          lung lobe morphogenesis
          lung epithelial cell differentiation
          epithelial cell morphogenesis involved in placental branching
          bicellular tight junction assembly
          epithelium migration
          intronic transcription regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          chromatin DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08008613.hg.1 ENST00000251808.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08008613.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08008613.hg.1 ENST00000395928.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08008613.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08008613.hg.1 TC08000629.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000521085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 ENST00000521085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800157879.hg.1 TC0800008416.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000251808.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000395927.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000472106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000474338.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000517674.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000521085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 ENST00000521085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq
          Affymetrix Human Clariom D Assay TC0800008416.hg.1 NM_024915.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline