Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01056452_m1
          See other PTK2 GE Assays ›
          Gene Symbol:
          PTK2
          Gene Name:
          protein tyrosine kinase 2
          Gene Aliases:
          FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
          Chromosome Location:
          Chr.8: 140658382 - 141001313 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01056452_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199649.1 NP_001186578.1 25 - 26 2677 105
          NM_001316342.1 NP_001303271.1 25 - 26 2460 105
          NM_005607.4 NP_005598.3 25 - 26 2666 105
          NM_153831.3 NP_722560.1 25 - 26 2677 105
          XM_005251003.2 XP_005251060.1 27 - 28 2841 105
          XM_005251012.2 XP_005251069.1 27 - 28 2683 105
          XM_005251013.2 XP_005251070.1 26 - 27 2502 105
          XM_006716608.2 XP_006716671.1 28 - 29 2772 105
          XM_006716609.2 XP_006716672.1 27 - 28 2605 105
          XM_006716610.2 XP_006716673.1 27 - 28 2597 105
          XM_011517194.2 XP_011515496.1 28 - 29 2741 105
          XM_017013651.1 XP_016869140.1 26 - 27 2522 105
          XM_017013652.1 XP_016869141.1 26 - 27 2522 105
          XM_017013653.1 XP_016869142.1 26 - 27 2757 105
          XM_017013654.1 XP_016869143.1 25 - 26 2739 105
          XM_017013655.1 XP_016869144.1 26 - 27 2733 105
          XM_017013656.1 XP_016869145.1 25 - 26 2739 105
          XM_017013657.1 XP_016869146.1 24 - 25 2718 105
          XM_017013658.1 XP_016869147.1 27 - 28 2644 105
          XM_017013659.1 XP_016869148.1 23 - 24 2399 105
          XM_017013660.1 XP_016869149.1 27 - 28 2581 105
          XM_017013661.1 XP_016869150.1 26 - 27 2599 105
          XM_017013662.1 XP_016869151.1 26 - 27 2563 105
          XM_017013663.1 XP_016869152.1 25 - 26 2494 105
          XM_017013664.1 XP_016869153.1 26 - 27 2667 105
          XM_017013665.1 XP_016869154.1 25 - 26 2491 105
          XM_017013666.1 XP_016869155.1 23 - 24 2610 105
          XM_017013667.1 XP_016869156.1 27 - 28 2557 105
          XM_017013668.1 XP_016869157.1 26 - 27 2575 105
          XM_017013669.1 XP_016869158.1 26 - 27 2563 105
          XM_017013670.1 XP_016869159.1 26 - 27 2560 105
          XM_017013671.1 XP_016869160.1 25 - 26 2542 105
          XM_017013672.1 XP_016869161.1 24 - 25 2473 105
          XM_017013673.1 XP_016869162.1 24 - 25 2473 105
          XM_017013674.1 XP_016869163.1 27 - 28 2688 105
          XM_017013675.1 XP_016869164.1 26 - 27 2512 105
          XM_017013676.1 XP_016869165.1 23 - 24 2399 105
          XM_017013677.1 XP_016869166.1 26 - 27 2473 105
          XM_017013678.1 XP_016869167.1 23 - 24 2365 105
          XM_017013679.1 XP_016869168.1 25 - 26 2404 105
          XM_017013680.1 XP_016869169.1 27 - 28 3657 105
          XM_017013681.1 XP_016869170.1 27 - 28 2605 105
          XM_017013682.1 XP_016869171.1 25 - 26 2481 105
          XM_017013683.1 XP_016869172.1 26 - 27 2521 105
          XM_017013684.1 XP_016869173.1 25 - 26 2503 105
          XM_017013685.1 XP_016869174.1 25 - 26 2444 105
          XM_017013686.1 XP_016869175.1 23 - 24 2379 105
          XM_017013687.1 XP_016869176.1 26 - 27 2521 105
          XM_017013688.1 XP_016869177.1 25 - 26 2413 105
          XM_017013689.1 XP_016869178.1 24 - 25 2278 105
          XM_017013690.1 XP_016869179.1 23 - 24 2234 105
          GenBank mRNA AB209071.1 - 25 - 26 2366 105
          AB209083.1 - 28 - 29 2387 105
          AK094999.1 - 15 - 16 1383 105
          AK294873.1 - 16 - 17 1461 105
          AK301557.1 - 4 - 5 382 105
          AK304356.1 - 25 - 26 2522 105
          AL832961.1 - 25 - 26 2549 105
          BC028733.2 - 11 - 12 1325 4812915 105
          KF437463.1 - 23 - 24 2370 105
          L13616.1 - 25 - 26 2599 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800012001.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001686.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTK2

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          microtubule cytoskeleton organization
          angiogenesis
          vasculogenesis
          neuron migration
          placenta development
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart morphogenesis
          cellular component disassembly involved in execution phase of apoptosis
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          axon guidance
          positive regulation of cell proliferation
          regulation of cell shape
          embryo development
          regulation of endothelial cell migration
          regulation of epithelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          central nervous system neuron axonogenesis
          negative regulation of cell-cell adhesion
          establishment of cell polarity
          extracellular matrix organization
          positive regulation of cell migration
          regulation of cell adhesion mediated by integrin
          netrin-activated signaling pathway
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          establishment of nucleus localization
          regulation of cell proliferation
          negative regulation of apoptotic process
          regulation of GTPase activity
          endothelial cell migration
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          regulation of osteoblast differentiation
          positive regulation of protein kinase activity
          negative regulation of organ growth
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          cell motility
          negative regulation of axonogenesis
          regulation of cytoskeleton organization
          regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of synapse assembly
          growth hormone receptor signaling pathway
          regulation of substrate adhesion-dependent cell spreading
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of anoikis
          actin binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          protein binding
          ATP binding
          JUN kinase binding
          protein kinase binding
          SH2 domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000340930.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000354438.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000395214.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000395218.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000395221.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000430260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000517887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000519419.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000519465.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000521059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000522684.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000535192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 ENST00000538769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 NM_001199649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 NM_005607.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 NM_153831.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08022095.hg.1 TC08001686.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000340930.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000395218.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000430260.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000517887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000518173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519419.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519654.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519993.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521562.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521986.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000522424.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000522684.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000523539.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000524202.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_001199649.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_005607.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_153831.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000340930.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000395218.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000430260.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000517887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000518173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519419.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519654.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000519993.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521562.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000521986.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000522424.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000522684.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000523539.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 ENST00000524202.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_001199649.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_005607.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 NM_153831.3 Refseq
          Affymetrix Human Clariom D Assay PSR0800181575.hg.1 TC0800012001.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline