| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018487.hg.1 |
ENST00000316386.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018487.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018487.hg.1 |
TC02001202.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174609.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174609.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174609.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174609.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174609.hg.1 |
TC0200010527.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000316386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
ENST00000435193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010527.hg.1 |
NM_012092.3 |
Refseq |