Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01059573_s1
          See other LOC51145 GE Assays ›
          Gene Symbol:
          LOC51145
          Gene Name:
          uncharacterized LOC51145
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 125743754 - 125745929 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01059573_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC51145
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF159054.1 - 1 1651 145

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitogen-activated protein kinase associated protein 1 MAPKAP1 JC310,
          MIP1,
          SIN1,
          SIN1b,
          SIN1g
          -37.1kb Sense Chr. 9 - 125437394 - 125707234 on NCBI Build GRCh38
          Downstream PBX homeobox 3 PBX3 786.0 Anti-sense Chr. 9 - 125747338 - 125967377 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27307233 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 17130 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 158197 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 11945 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 26761 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 411828 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 66719 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P115529 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 453 - -
          Agilent Human Genome CGH Microarray 44B A_14_P117320 - -
          Agilent Human Genome CGH Microarray 44B A_14_P202316 - -
          Agilent Human Genome CGH Microarray 44A A_14_P117320 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38236 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34618 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3404281 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P115529 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00319331 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P115529 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P202316 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P117320 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P202316 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P117320 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 104302 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 179445 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 29597 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 15704 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P117320 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P202316 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P117320 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P202316 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8164197 - -
          Affymetrix Human Gene 1.0 ST Array 8164199 - -
          Affymetrix Human Gene 1.0 ST Array 8164198 - -
          Affymetrix Human Gene 1.1 ST Array 8164196 - -
          Affymetrix HT HG-U133+ PM Array Plate 220752_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000444800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000444800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 NR_122032.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 NR_122032.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011536.hg.1 NR_122032.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000444800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000444800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000606827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 ENST00000614664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 NR_122032.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 NR_122032.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011536.hg.1 NR_122032.1 Refseq
          Human Genome U133 Plus 2.0 Array 220752_at - -
          Human Genome U133 Plus 2.0 Array 220752_PM_at - -
          Human Genome U133A 2.0 Array 220752_PM_at - -
          Human Genome U133A 2.0 Array 220752_at - -
          AffymetrixPathwork Diagnostics Pathchip pw522487 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220752_at4 - -
          Human Genome U95 Set 42385_at - -
          Human Genome U95 Set 42385_PM_at - -
          Human Genome U133 Set 220752_PM_at - -
          Human Genome U133 Set 220752_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 189265 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1682607 AF159054.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1682607 AF159054.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1682607 AF159054.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1682607 AF159054.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1682607 AF159054.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline