| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022368.hg.1 |
ENST00000281156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022368.hg.1 |
ENST00000539571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022368.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022368.hg.1 |
TC06001842.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022369.hg.1 |
ENST00000281156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022369.hg.1 |
ENST00000539571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022369.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022369.hg.1 |
TC06001842.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
ENST00000511849.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012175.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183916.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183916.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183916.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183916.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183916.hg.1 |
TC0600012175.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600183918.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183918.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183918.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183918.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183918.hg.1 |
TC0600012175.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600183919.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183919.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183919.hg.1 |
TC0600012175.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600183917.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183917.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183917.hg.1 |
ENST00000281156.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183917.hg.1 |
NM_152688.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183917.hg.1 |
TC0600012175.hg.1 |
Clariom D TC |