Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01061533_m1
          See other TMCO4 GE Assays ›
          Gene Symbol:
          TMCO4
          Gene Name:
          transmembrane and coiled-coil domains 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 19682213 - 19800385 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01061533_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMCO4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181719.4 NP_859070.3 10 - 11 1118 118
          XM_005245822.4 XP_005245879.1 10 - 11 1134 118
          XM_005245824.4 XP_005245881.1 10 - 11 1198 118
          XM_005245825.4 XP_005245882.1 9 - 10 999 118
          XM_006710551.3 XP_006710614.1 10 - 11 1223 118
          XM_011541179.2 XP_011539481.1 13 - 14 1593 118
          XM_011541180.2 XP_011539482.1 13 - 14 1748 118
          XM_011541181.1 XP_011539483.1 12 - 13 1422 118
          XM_011541185.2 XP_011539487.1 10 - 11 1141 118
          XM_011541186.2 XP_011539488.1 10 - 11 1238 118
          XM_017000914.1 XP_016856403.1 12 - 13 1642 118
          XM_017000915.1 XP_016856404.1 12 - 13 1364 118
          XM_017000916.1 XP_016856405.1 10 - 11 1276 118
          XM_017000917.1 XP_016856406.1 11 - 12 1491 118
          XM_017000918.1 XP_016856407.1 12 - 13 1688 118
          XM_017000919.1 XP_016856408.1 11 - 12 1415 118
          XM_017000920.1 XP_016856409.1 10 - 11 1633 118
          XM_017000921.1 XP_016856410.1 11 - 12 1440 118
          XM_017000922.1 XP_016856411.1 9 - 10 1099 118
          GenBank mRNA AL049846.1 - 7 - 8 884 118
          AX775875.1 - 9 - 10 1147 118
          BC053600.1 - 8 - 9 977 5744211 118
          BX640987.1 - 10 - 11 1119 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013153.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002304.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TMCO4

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01035298.hg.1 ENST00000294543.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035298.hg.1 ENST00000375122.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035298.hg.1 ENST00000375127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035298.hg.1 NM_181719.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035298.hg.1 TC01002304.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000294543.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000375127.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000489135.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 NM_181719.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000294543.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000375127.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 ENST00000489135.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 NM_181719.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100197336.hg.1 TC0100013153.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline