Search
Search

SUMF1| Gene Symbol: SUMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164674.1 | NP_001158146.1 | 2 - 3 | 488 | 117 | |
| XM_017006253.1 | XP_016861742.1 | 2 - 3 | 476 | 117 | ||
| GenBank mRNA | AK302018.1 | - | 2 - 3 | 456 | 117 | |
Set Membership: |
|