Search
Search

FAM207A| Gene Symbol: FAM207A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316983.1 | NP_001303912.1 | 2 - 3 | 344 | 112 | |
| NM_001316984.1 | NP_001303913.1 | 2 - 3 | 67 | |||
| NM_001316985.1 | NP_001303914.1 | 2 - 3 | 67 | |||
| NM_001316986.1 | NP_001303915.1 | 2 - 3 | 344 | 112 | ||
| NM_001316987.1 | NP_001303916.1 | 2 - 3 | 67 | |||
| NM_001316988.1 | NP_001303917.1 | 2 - 3 | 67 | |||
| NM_058190.3 | NP_478070.1 | 2 - 3 | 344 | 112 | ||
| XM_017028481.1 | XP_016883970.1 | 2 - 3 | 492 | 112 | ||
| GenBank mRNA | AF391113.1 | - | 2 - 3 | 325 | 112 | |
| AF391114.1 | - | 2 - 3 | 67 | |||
| BC009341.2 | - | 2 - 3 | 301 | 4128732 | 112 | |
| BG479572.1 | - | 2 - 3 | 261 | 112 | ||
| BG744869.1 | - | 4 - 5 | 67 | |||
| BM042090.1 | - | 2 - 3 | 67 | |||
| CN368049.1 | - | 2 - 3 | 332 | 112 | ||
| CR998572.1 | - | 2 - 3 | 67 | |||
| HY097926.1 | - | 2 - 3 | 67 | |||
Set Membership: |
|