| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000518699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
ENST00000524115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010251.hg.1 |
NM_152568.3 |
Refseq |