Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01063538_m1
          See other ERCC4 GE Assays ›
          Gene Symbol:
          ERCC4
          Gene Name:
          ERCC excision repair 4, endonuclease catalytic subunit
          Gene Aliases:
          ERCC11, FANCQ, RAD1, XFEPS, XPF
          Chromosome Location:
          Chr.16: 13920138 - 13952348 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01063538_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERCC4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005236.2 NP_005227.1 9 - 10 1910 92
          XM_011522424.2 XP_011520726.1 10 - 11 2067 92
          XM_011522427.1 XP_011520729.1 4 - 5 1047 92
          XM_017023043.1 XP_016878532.1 10 - 11 2024 92
          GenBank mRNA AK289726.1 - 9 - 10 1909 92
          AK301930.1 - 4 - 5 1179 92
          U64315.1 - 9 - 10 1883 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600006976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000170.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ERCC4

          Panther Classification:

          Molecular Function -

          DNA metabolism protein endodeoxyribonuclease nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          resolution of meiotic recombination intermediates
          telomere maintenance
          double-strand break repair via homologous recombination
          DNA repair
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          response to UV
          UV protection
          negative regulation of telomere maintenance
          nucleotide-excision repair, DNA incision
          cellular response to UV
          interstrand cross-link repair
          global genome nucleotide-excision repair
          nucleotide-excision repair involved in interstrand cross-link repair
          single-stranded DNA endodeoxyribonuclease activity
          TFIID-class transcription factor binding
          damaged DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          protein binding
          protein C-terminus binding
          structure-specific DNA binding
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16002588.hg.1 ENST00000311895.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002588.hg.1 ENST00000389138.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002588.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002588.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16002588.hg.1 TC16000170.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000573018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574194.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000574781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000575156.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 ENST00000576348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006976.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 NM_005236.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 TC1600006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000311895.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000389138.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 ENST00000462862.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600149417.hg.1 NM_005236.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 167294 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline