| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05016814.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05016814.hg.1 |
ENST00000503939.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05016814.hg.1 |
ENST00000535119.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05016814.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05016814.hg.1 |
TC05001172.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500171218.hg.1 |
TC0500010169.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000284268.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000502585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000503939.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000505140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000513115.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
ENST00000515517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010169.hg.1 |
NM_054027.5 |
Refseq |