Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01065107_g1
          See other FCHO1 GE Assays ›
          Gene Symbol:
          FCHO1
          Gene Name:
          FCH domain only 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 17747718 - 17788568 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01065107_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FCHO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161357.1 NP_001154829.1 17 - 18 1487 83
          NM_001161358.1 NP_001154830.1 16 - 17 1397 83
          NM_001161359.1 NP_001154831.1 15 - 16 1327 83
          NM_015122.2 NP_055937.1 17 - 18 1491 83
          XM_005259831.3 XP_005259888.1 16 - 17 1576 83
          XM_006722701.3 XP_006722764.1 16 - 17 1495 83
          XM_006722702.3 XP_006722765.1 16 - 17 1572 83
          XM_011527824.2 XP_011526126.1 17 - 18 1487 83
          XM_011527825.2 XP_011526127.1 17 - 18 1476 83
          XM_011527826.2 XP_011526128.1 17 - 18 1459 83
          XM_011527827.2 XP_011526129.1 15 - 16 1294 83
          XM_011527828.2 XP_011526130.1 16 - 17 1390 83
          XM_017026519.1 XP_016882008.1 17 - 18 1557 83
          XM_017026520.1 XP_016882009.1 17 - 18 1540 83
          XM_017026521.1 XP_016882010.1 16 - 17 1491 83
          XM_017026522.1 XP_016882011.1 17 - 18 1487 83
          XM_017026523.1 XP_016882012.1 17 - 18 1557 83
          XM_017026524.1 XP_016882013.1 16 - 17 1572 83
          XM_017026525.1 XP_016882014.1 16 - 17 1370 83
          XM_017026526.1 XP_016882015.1 17 - 18 1487 83
          XM_017026527.1 XP_016882016.1 16 - 17 1495 83
          XM_017026528.1 XP_016882017.1 17 - 18 1557 83
          GenBank mRNA AB006628.1 - 14 - 15 1261 83
          AK291410.1 - 16 - 17 1353 83
          AK303623.1 - 13 - 14 1189 83
          AK316458.1 - 15 - 16 1298 83
          BC028021.1 - 17 - 18 1432 5218319 83
          BC041130.1 - 17 - 18 1441 5757146 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007342.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000342.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FCHO1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          clathrin coat assembly
          clathrin-mediated endocytosis
          protein binding
          AP-2 adaptor complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 ENST00000252771.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 ENST00000389133.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 ENST00000539407.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 NM_001161357.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 NM_001161358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 NM_001161359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 NM_015122.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004532.hg.1 TC19000342.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000252771.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000594202.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000595033.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000596536.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000596951.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000597512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000599766.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000600676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000602111.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161357.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161358.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_015122.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 TC1900007342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000252771.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000594202.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000595033.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000596536.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000596951.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000597512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000599766.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000600676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 ENST00000602111.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161357.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161358.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_001161359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900153717.hg.1 NM_015122.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline