Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01066930_m1
          See other MDM2 GE Assays ›
          Gene Symbol:
          MDM2
          Gene Name:
          MDM2 proto-oncogene
          Gene Aliases:
          ACTFS, HDMX, LSKB, hdm2
          Chromosome Location:
          Chr.12: 68808172 - 68850686 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01066930_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MDM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145337.3 NP_001138809.1 2 - 3 146 99
          NM_001145339.2 NP_001138811.1 2 - 3 403 99
          NM_001145340.3 NP_001138812.1 2 - 3 146 99
          NM_001278462.2 NP_001265391.1 2 - 3 146 99
          NM_001367990.1 NP_001354919.1 2 - 3 153 99
          NM_002392.6 NP_002383.2 2 - 3 398 99
          GenBank mRNA AF092843.1 - 2 - 3 111 99
          AF092844.1 - 2 - 3 111 99
          AF092845.1 - 2 - 3 111 99
          AF385322.1 - 1 - 2 79 99
          AF385323.1 - 1 - 2 79 99
          AF385324.1 - 1 - 2 79 99
          AF385326.1 - 1 - 2 79 99
          AF385327.1 - 1 - 2 79 99
          AJ276888.1 - 1 - 2 79 99
          AJ278975.1 - 1 - 2 79 99
          AJ278977.1 - 1 - 2 79 99
          AJ430612.1 - 1 - 2 79 99
          AJ430614.1 - 1 - 2 79 99
          AJ491698.1 - 1 - 2 79 99
          AJ491699.1 - 1 - 2 79 99
          AJ491700.1 - 1 - 2 79 99
          AJ491701.1 - 1 - 2 79 99
          AJ550517.1 - 1 - 2 79 99
          AJ550518.1 - 1 - 2 79 99
          AJ550519.1 - 1 - 2 79 99
          AK290341.1 - 2 - 3 353 99
          AM902642.1 - 1 - 2 79 99
          AM902643.1 - 1 - 2 79 99
          AM902644.1 - 1 - 2 79 99
          AM902646.1 - 1 - 2 79 99
          AM902647.1 - 1 - 2 79 99
          AM902648.1 - 1 - 2 79 99
          AM902649.1 - 1 - 2 79 99
          AM902650.1 - 1 - 2 79 99
          AM902651.1 - 1 - 2 79 99
          AM902654.1 - 1 - 2 79 99
          AM902655.1 - 1 - 2 79 99
          AM902656.1 - 1 - 2 79 99
          AM902657.1 - 1 - 2 79 99
          AM902658.1 - 1 - 2 79 99
          AM902659.1 - 1 - 2 79 99
          AM902660.1 - 1 - 2 79 99
          AM902661.1 - 1 - 2 79 99
          AM902663.1 - 1 - 2 79 99
          AM902664.1 - 1 - 2 79 99
          AM902665.1 - 1 - 2 79 99
          AM902666.1 - 1 - 2 79 99
          AM902667.1 - 1 - 2 79 99
          AM902668.1 - 1 - 2 79 99
          AM902669.1 - 1 - 2 79 99
          AM902672.1 - 1 - 2 79 99
          AM902674.1 - 1 - 2 79 99
          AM902675.1 - 1 - 2 79 99
          AM902678.1 - 1 - 2 79 99
          AM902679.1 - 1 - 2 79 99
          AM902680.1 - 1 - 2 79 99
          AM902681.1 - 1 - 2 79 99
          BC152384.1 - 1 - 2 82 99
          BC152390.1 - 1 - 2 82 99
          BE296905.1 - 2 - 3 368 99
          BT007258.1 - 1 - 2 79 99
          EU076747.1 - 2 - 3 99
          EU076749.1 - 2 - 3 153 99
          HM213901.1 - 1 - 2 79 99
          HM213902.1 - 1 - 2 79 99
          HM213903.1 - 1 - 2 79 99
          HM213904.1 - 1 - 2 79 99
          HY174841.1 - 2 - 3 405 99
          JX015379.1 - 1 - 2 79 99
          JX015380.1 - 1 - 2 79 99
          JX015382.1 - 1 - 2 79 99
          JX015383.1 - 1 - 2 79 99
          JX015384.1 - 1 - 2 79 99
          JX015385.1 - 1 - 2 79 99
          JX015387.1 - 1 - 2 79 99
          JX015388.1 - 1 - 2 79 99
          JX015389.1 - 1 - 2 79 99
          JX015390.1 - 1 - 2 79 99
          JX015391.1 - 1 - 2 79 99
          JX015392.1 - 1 - 2 79 99
          M92424.1 - 2 - 3 390 99
          U33201.1 - 1 - 2 79 99
          U33203.1 - 1 - 2 79 99
          Z12020.1 - 2 - 3 390 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008116.hg
          This assay targets the same location as HTA transcript cluster(s) TC12000606.hg

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MDM2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein polyubiquitination
          blood vessel development
          blood vessel remodeling
          regulation of heart rate
          heart valve development
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          ventricular septum development
          atrial septum development
          ubiquitin-dependent protein catabolic process
          apoptotic process
          traversing start control point of mitotic cell cycle
          heart development
          positive regulation of cell proliferation
          response to xenobiotic stimulus
          response to toxic substance
          response to iron ion
          regulation of gene expression
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of protein processing
          negative regulation of neuron projection development
          protein ubiquitination
          protein sumoylation
          DNA damage response, signal transduction by p53 class mediator
          protein destabilization
          response to magnesium ion
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein localization to nucleus
          regulation of protein catabolic process
          response to cocaine
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          protein-containing complex subunit organization
          establishment of protein localization
          response to ether
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          response to antibiotic
          positive regulation of protein export from nucleus
          response to steroid hormone
          positive regulation of muscle cell differentiation
          proteolysis involved in cellular protein catabolic process
          regulation of cell cycle
          protein autoubiquitination
          cardiac septum morphogenesis
          protein-containing complex assembly
          regulation of biological quality
          cellular response to hydrogen peroxide
          cellular response to antibiotic
          cellular response to vitamin B1
          cellular response to alkaloid
          cellular response to growth factor stimulus
          cellular response to peptide hormone stimulus
          cellular response to estrogen stimulus
          cellular response to hypoxia
          cellular response to gamma radiation
          cellular response to UV-C
          signal transduction by p53 class mediator
          fibroblast activation
          cellular response to actinomycin D
          regulation of postsynaptic neurotransmitter receptor internalization
          regulation of protein catabolic process at postsynapse, modulating synaptic transmission
          negative regulation of signal transduction by p53 class mediator
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          response to formaldehyde
          positive regulation of vascular smooth muscle cell proliferation
          positive regulation of vascular associated smooth muscle cell migration
          amyloid fibril formation
          response to water-immersion restraint stress
          negative regulation of apoptotic signaling pathway
          p53 binding
          ubiquitin-protein transferase activity
          protein binding
          5S rRNA binding
          zinc ion binding
          transferase activity
          ligase activity
          SUMO transferase activity
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          receptor serine/threonine kinase binding
          identical protein binding
          peroxisome proliferator activated receptor binding
          ribonucleoprotein complex binding
          ubiquitin binding
          metal ion binding
          ubiquitin protein ligase activity
          NEDD8 ligase activity
          disordered domain specific binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000258148.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000258149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000299252.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000311420.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000311440.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000356290.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000358483.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000393410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000462284.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000517852.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000544561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 ENST00000544648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 NM_002392.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12008008.hg.1 TC12000606.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000356290.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000393415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000393416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000523991.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000539479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000543323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001145337.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001145340.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001278462.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000356290.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000393415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000393416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000523991.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000539479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 ENST00000543323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001145337.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001145340.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 NM_001278462.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159426.hg.1 TC1200008116.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline